siRNA Details
| Gene Symbol: Lrch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033439.3 | NP_001028611.2 | 10, 11 | 1493 | ||
| NM_001252132.1 | NP_001239061.1 | 10, 11 | 1493 | |||
| GenBank mRNA | AK129264.1 | - | 10, 11 | 1482 | ||
| AK136026.1 | - | 10, 11 | 1099 | |||
| AK143909.1 | - | 10, 11 | 1492 | |||
| AK160902.1 | - | 8, 9 | 869 | |||

Lrch1
Genomic Map