siRNA Details
| Gene Symbol: Lmo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201529.2 | NP_963287.2 | 30 | 5270 | ||
| GenBank mRNA | AK129231.1 | - | 26 | 4946 | ||
| AK137694.1 | - | 21 | 3615 | |||
| BC082553.1 | - | 27 | 5176 | |||
| BC145011.1 | - | 23 | 3754 | 9053559 | ||
| EF071857.1 | - | 30 | 5241 | |||

Lmo7
Genomic Map