siRNA Details
| Gene Symbol: Vps13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177151.3 | NP_796125.2 | 30, 31 | 4992 | ||
| GenBank mRNA | BQ558426.1 | - | 1, 2 | 16 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps13b| Gene Symbol: Vps13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177151.3 | NP_796125.2 | 30, 31 | 4992 | ||
| GenBank mRNA | BQ558426.1 | - | 1, 2 | 16 | ||