siRNA Details
| Gene Symbol: Zfat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145888.1 | NP_001139360.1 | 6 | 1936 | ||
| NM_198644.2 | NP_941046.1 | 6 | 1915 | |||
| GenBank mRNA | BC053930.1 | - | 6 | 1906 | 6389756 | |
| EU221277.1 | - | 6 | 1766 | |||

Zfat
Genomic Map