siRNA Details
Gene Symbol: Cdkl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033443.4 | NP_001028615.1 | 4 | 500 | ||
GenBank mRNA | AK157995.1 | - | 4 | 501 | ||
BC138986.1 | - | 3 | 398 | 8862008 | ||
BC138988.1 | - | 3 | 398 | 8862010 |
