siRNA Details
| Gene Symbol: Skor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109743.1 | NP_001103213.1 | 2 | 597 | ||
| GenBank mRNA | AB358976.1 | - | 2 | 597 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Skor2| Gene Symbol: Skor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109743.1 | NP_001103213.1 | 2 | 597 | ||
| GenBank mRNA | AB358976.1 | - | 2 | 597 | ||