siRNA Details
| Gene Symbol: Greb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083628.1 | NP_001077097.1 | 29 | 5252 | ||
| GenBank mRNA | AK220484.1 | - | 9 | 1769 | ||
| BC147570.1 | - | 28 | 5109 | 9056560 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Greb1l| Gene Symbol: Greb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083628.1 | NP_001077097.1 | 29 | 5252 | ||
| GenBank mRNA | AK220484.1 | - | 9 | 1769 | ||
| BC147570.1 | - | 28 | 5109 | 9056560 | ||