siRNA Details
Gene Symbol: Gm960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033447.3 | NP_001028619.2 | 5 | 820 | ||
GenBank mRNA | AK134945.1 | - | 5 | 468 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033447.3 | NP_001028619.2 | 5 | 820 | ||
GenBank mRNA | AK134945.1 | - | 5 | 468 |