siRNA Details
Gene Symbol: Fam189a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114174.1 | NP_001107646.1 | 5 | 862 | ||
GenBank mRNA | DV650276.1 | - | 3 | 261 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam189a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114174.1 | NP_001107646.1 | 5 | 862 | ||
GenBank mRNA | DV650276.1 | - | 3 | 261 |