siRNA Details
Gene Symbol: 4430402I18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198651.2 | NP_941053.2 | 10 | 1156 | ||
GenBank mRNA | AK031997.1 | - | 10 | 1156 | ||
BC147044.1 | - | 9 | 736 | 9056033 | ||
BC147045.1 | - | 9 | 736 | 9056045 | ||
BC171985.1 | - | 8 | 685 | 9054271 | ||
BC171986.1 | - | 8 | 694 | 9054272 |
