siRNA Details
| Gene Symbol: Atp2b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167949.2 | NP_001161421.1 | 8 | 1705 | ||
| NM_213616.4 | NP_998781.1 | 8 | 1705 | |||
| GenBank mRNA | AY560895.1 | - | 8 | 1239 | ||
| BC109172.1 | - | 8 | 1239 | 40048499 | ||
| BC109173.1 | - | 8 | 1239 | 40048500 | ||
| EU493094.2 | - | 8 | 1409 | |||
| EU493095.2 | - | 8 | 1409 | |||
| JF683384.1 | - | 8 | 1409 | |||

Atp2b4
Genomic Map