siRNA Details
| Gene Symbol: Axdnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK134071.1 | - | 3 | 261 | ||
| BC150932.1 | - | 2 | 113 | 9087843 | ||
| BC172088.1 | - | 2 | 113 | 9054374 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Axdnd1| Gene Symbol: Axdnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK134071.1 | - | 3 | 261 | ||
| BC150932.1 | - | 2 | 113 | 9087843 | ||
| BC172088.1 | - | 2 | 113 | 9054374 | ||