siRNA Details
| Gene Symbol: Lonrf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029878.1 | NP_001025049.1 | 4 | 517 | ||
| GenBank mRNA | BC139127.1 | - | 4 | 568 | 8862149 | |
| BC139130.1 | - | 4 | 568 | 8862152 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lonrf2| Gene Symbol: Lonrf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029878.1 | NP_001025049.1 | 4 | 517 | ||
| GenBank mRNA | BC139127.1 | - | 4 | 568 | 8862149 | |
| BC139130.1 | - | 4 | 568 | 8862152 | ||