siRNA Details
Gene Symbol: Tmem182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081198.1 | NP_001074667.1 | 3 | 410 | ||
GenBank mRNA | BC147440.1 | - | 3 | 367 | 9056082 | |
BC147458.1 | - | 3 | 367 | 9056100 | ||
BC147740.1 | - | 3 | 367 | 9056689 | ||
BC147743.1 | - | 3 | 367 | 9056692 |
