siRNA Details
| Gene Symbol: Tmem182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081198.1 | NP_001074667.1 | 1 | 246 | ||
| GenBank mRNA | BC147440.1 | - | 1 | 203 | 9056082 | |
| BC147458.1 | - | 1 | 203 | 9056100 | ||
| BC147740.1 | - | 1 | 203 | 9056689 | ||
| BC147743.1 | - | 1 | 203 | 9056692 | ||

Tmem182
Genomic Map