siRNA Details
| Gene Symbol: Sptan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177667.1 | NP_001171138.1 | 10 | 1526 | ||
| NM_001177668.1 | NP_001171139.1 | 10 | 1526 | |||
| GenBank mRNA | AK037683.1 | - | 7 | 3145 | ||
| AK132224.1 | - | 10 | 1410 | |||
| AK141149.1 | - | 10 | 1407 | |||
| AK147317.1 | - | 10 | 1410 | |||
| BC150941.1 | - | 10 | 1362 | 9087852 | ||
| BC158015.1 | - | 10 | 1362 | 9007321 | ||
| BC172095.1 | - | 10 | 1362 | 9054381 | ||

Sptan1
Genomic Map