siRNA Details
| Gene Symbol: Med19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025885.3 | NP_080161.1 | 2 | 289 | ||
| GenBank mRNA | AK005399.1 | - | 2 | 257 | ||
| AK010552.1 | - | 2 | 267 | |||
| AK160579.1 | - | 2 | 255 | |||
| AK168787.1 | - | 2 | 257 | |||
| AK209282.1 | - | 2 | 57 | |||
| BB611514.1 | - | 2 | 289 | |||
| BC044207.1 | - | 2 | 151 | |||
| BC044302.1 | - | 2 | 151 | |||
| BC055404.1 | - | 2 | 244 | |||
| BC082559.1 | - | 2 | 254 | 30606373 | ||

Med19
Genomic Map