siRNA Details
| Gene Symbol: Cdh26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291189.1 | NP_001278118.1 | 14 | 2346 | ||
| NM_198656.2 | NP_941058.1 | 15 | 2403 | |||
| GenBank mRNA | AK132328.1 | - | 14 | 2331 | ||
| BC053928.1 | - | 15 | 2311 | 6337144 | ||

Cdh26
Genomic Map