siRNA Details
| Gene Symbol: Stpg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198659.2 | NP_941061.1 | 1, 2 | 118 | ||
| GenBank mRNA | AK046955.1 | - | 1, 2 | 119 | ||
| AK161389.1 | - | 1, 2 | 113 | |||
| BC138108.1 | - | 1, 2 | 157 | 8861127 | ||
| BC138109.1 | - | 1, 2 | 157 | 8861128 | ||

Stpg2
Genomic Map