siRNA Details
| Gene Symbol: Rxfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212452.1 | NP_997617.1 | 18 | 2137 | ||
| GenBank mRNA | AK137655.1 | - | 16 | 4240 | ||
| AY509975.1 | - | 18 | 2137 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rxfp1| Gene Symbol: Rxfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212452.1 | NP_997617.1 | 18 | 2137 | ||
| GenBank mRNA | AK137655.1 | - | 16 | 4240 | ||
| AY509975.1 | - | 18 | 2137 | |||