siRNA Details
| Gene Symbol: C87499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198663.3 | NP_941065.2 | 4 | 1453 | ||
| GenBank mRNA | AK135883.1 | - | 4 | 1453 | ||
| AK163216.1 | - | 3 | 1144 | |||
| BC053719.1 | - | 4 | 1454 | 30053512 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C87499| Gene Symbol: C87499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198663.3 | NP_941065.2 | 4 | 1453 | ||
| GenBank mRNA | AK135883.1 | - | 4 | 1453 | ||
| AK163216.1 | - | 3 | 1144 | |||
| BC053719.1 | - | 4 | 1454 | 30053512 | ||