siRNA Details
| Gene Symbol: Rbm33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028234.1 | NP_082510.1 | 5 | 467 | ||
| GenBank mRNA | AK014289.1 | - | 5 | 456 | ||
| BB637766.1 | - | 5 | 480 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm33| Gene Symbol: Rbm33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028234.1 | NP_082510.1 | 5 | 467 | ||
| GenBank mRNA | AK014289.1 | - | 5 | 456 | ||
| BB637766.1 | - | 5 | 480 | |||