siRNA Details
| Gene Symbol: B3glct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081204.1 | NP_001074673.1 | 13, 14 | 1247 | ||
| GenBank mRNA | AB253762.1 | - | 13, 14 | 1212 | ||
| AK145082.1 | - | 13, 14 | 1222 | |||
| BC115781.1 | - | 1 | 116 | |||
| BC117522.1 | - | 1 | 116 | |||

B3glct
Genomic Map