siRNA Details
| Gene Symbol: Drc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033460.3 | NP_001028632.1 | 598 | |||
| GenBank mRNA | AK140154.1 | - | 599 | |||
| BC150863.1 | - | 596 | 9087774 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Drc1| Gene Symbol: Drc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033460.3 | NP_001028632.1 | 598 | |||
| GenBank mRNA | AK140154.1 | - | 599 | |||
| BC150863.1 | - | 596 | 9087774 | |||