siRNA Details
| Gene Symbol: Wee2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201370.2 | NP_958758.2 | 8 | 1444 | ||
| GenBank mRNA | AK139573.1 | - | 8 | 1403 | ||
| AK143336.1 | - | 8 | 1368 | |||
| AK163344.1 | - | 8 | 1446 | |||
| BC052883.1 | - | 8 | 1351 | 30053490 | ||
| BC080784.1 | - | 8 | 1369 | 30481521 | ||
| DQ011691.1 | - | 8 | 1265 | |||

Wee2
Genomic Map