siRNA Details
| Gene Symbol: Wee2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201370.2 | NP_958758.2 | 2 | 395 | ||
| GenBank mRNA | AK139573.1 | - | 2 | 354 | ||
| AK143336.1 | - | 2 | 319 | |||
| AK163344.1 | - | 2 | 397 | |||
| BC052883.1 | - | 2 | 302 | 30053490 | ||
| BC080784.1 | - | 2 | 320 | 30481521 | ||
| BY725757.1 | - | 2 | 396 | |||
| DQ011691.1 | - | 2 | 216 | |||

Wee2
Genomic Map