siRNA Details
| Gene Symbol: Gm1698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133405.1 | - | 3 | 379 | ||
| BC172080.1 | - | 3 | 351 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm1698| Gene Symbol: Gm1698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133405.1 | - | 3 | 379 | ||
| BC172080.1 | - | 3 | 351 | |||