siRNA Details
| Gene Symbol: Odf3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198673.2 | NP_941075.1 | 3 | 314 | ||
| GenBank mRNA | AK142219.1 | - | 4 | 449 | ||
| BC049697.1 | - | 3 | 314 | 6706514 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Odf3l1| Gene Symbol: Odf3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198673.2 | NP_941075.1 | 3 | 314 | ||
| GenBank mRNA | AK142219.1 | - | 4 | 449 | ||
| BC049697.1 | - | 3 | 314 | 6706514 | ||