siRNA Details
| Gene Symbol: Gm1141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133067.1 | - | 5 | 878 | ||
| BC147557.1 | - | 4 | 733 | 9087440 | ||
| BC150928.1 | - | 4 | 733 | 9087839 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm1141| Gene Symbol: Gm1141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133067.1 | - | 5 | 878 | ||
| BC147557.1 | - | 4 | 733 | 9087440 | ||
| BC150928.1 | - | 4 | 733 | 9087839 | ||