siRNA Details
| Gene Symbol: Brwd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081477.1 | NP_001074946.1 | 7 | 724 | ||
| GenBank mRNA | AK042444.1 | - | 716 | |||
| AK050539.1 | - | 7 | 710 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brwd3| Gene Symbol: Brwd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081477.1 | NP_001074946.1 | 7 | 724 | ||
| GenBank mRNA | AK042444.1 | - | 716 | |||
| AK050539.1 | - | 7 | 710 | |||