siRNA Details
| Gene Symbol: Poc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027740.6 | NP_082016.1 | 7 | 825 | ||
| GenBank mRNA | AK016991.1 | - | 6 | 749 | ||
| AK030180.1 | - | 6 | 719 | |||
| AK045302.1 | - | 7 | 804 | |||
| AK050233.1 | - | 7 | 894 | |||
| AK090075.1 | - | 7 | 828 | |||
| BC050888.1 | - | 6 | 823 | |||
| BC099926.1 | - | 7 | 912 | |||
| BC144819.1 | - | 7 | 914 | |||
| BC145801.1 | - | 7 | 914 | 40131104 | ||

Poc1b
Genomic Map