siRNA Details
| Gene Symbol: Tmed8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033475.3 | NP_001028647.1 | 5 | 572 | ||
| GenBank mRNA | AK147378.1 | - | 5 | 573 | ||
| BC150860.1 | - | 5 | 643 | 9087771 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmed8| Gene Symbol: Tmed8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033475.3 | NP_001028647.1 | 5 | 572 | ||
| GenBank mRNA | AK147378.1 | - | 5 | 573 | ||
| BC150860.1 | - | 5 | 643 | 9087771 | ||