siRNA Details
Gene Symbol: Zfp384 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252083.1 | NP_001239012.1 | 6 | 987 | ||
NM_175557.5 | NP_780766.3 | 6 | 958 | |||
GenBank mRNA | AK086185.1 | - | 6 | 883 | ||
AK156026.1 | - | 6 | 989 | |||
AK172476.1 | - | 4 | 1377 | |||
BC064013.1 | - | 6 | 822 | 6515698 | ||
DQ323640.1 | - | 1 | 217 | |||
DQ323641.1 | - | 1 | 217 |
