siRNA Details
| Gene Symbol: Wdfy4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146022.2 | NP_001139494.1 | 3 | 472 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdfy4| Gene Symbol: Wdfy4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146022.2 | NP_001139494.1 | 3 | 472 | ||