siRNA Details
| Gene Symbol: Zfp488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013777.2 | NP_001013799.2 | 2 | 1076 | ||
| GenBank mRNA | BC089025.1 | - | 2 | 1076 | 30618054 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp488| Gene Symbol: Zfp488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013777.2 | NP_001013799.2 | 2 | 1076 | ||
| GenBank mRNA | BC089025.1 | - | 2 | 1076 | 30618054 | |