siRNA Details
| Gene Symbol: Fam188b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142781.1 | NP_001136253.1 | 15 | 1958 | ||
| GenBank mRNA | AK142385.1 | - | 15 | 1876 | ||
| AK161304.1 | - | 15 | 1837 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam188b| Gene Symbol: Fam188b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142781.1 | NP_001136253.1 | 15 | 1958 | ||
| GenBank mRNA | AK142385.1 | - | 15 | 1876 | ||
| AK161304.1 | - | 15 | 1837 | |||