siRNA Details
| Gene Symbol: Irs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081212.1 | NP_001074681.1 | 1 | 3713 | ||
| GenBank mRNA | AK155277.1 | - | 1 | 3535 | ||
| BC147580.1 | - | 1 | 3741 | 9056570 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irs2| Gene Symbol: Irs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081212.1 | NP_001074681.1 | 1 | 3713 | ||
| GenBank mRNA | AK155277.1 | - | 1 | 3535 | ||
| BC147580.1 | - | 1 | 3741 | 9056570 | ||