siRNA Details
| Gene Symbol: Adam20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009548.2 | NP_001009548.1 | 2 | 2279 | ||
| GenBank mRNA | AK133113.1 | - | 2 | 2280 | ||
| BC150793.1 | - | 2165 | 9087704 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam20| Gene Symbol: Adam20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009548.2 | NP_001009548.1 | 2 | 2279 | ||
| GenBank mRNA | AK133113.1 | - | 2 | 2280 | ||
| BC150793.1 | - | 2165 | 9087704 | |||