siRNA Details
| Gene Symbol: Pglyrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165968.1 | NP_001159440.1 | 6 | 694 | ||
| NM_207263.2 | NP_997146.2 | 6 | 691 | |||
| GenBank mRNA | AY518699.1 | - | 6 | 645 | ||
| BC120840.1 | - | 6 | 686 | 40129763 | ||
| BC144902.1 | - | 6 | 694 | 9053450 | ||
| BC144927.1 | - | 5 | 575 | |||

Pglyrp4
Genomic Map