siRNA Details
| Gene Symbol: Lca5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001492.2 | NP_001001492.2 | 4 | 573 | ||
| GenBank mRNA | AK029554.1 | - | 4 | 567 | ||
| AK133316.1 | - | 4 | 573 | |||
| BC141285.1 | - | 3 | 423 | 9055897 | ||
| BC145611.1 | - | 3 | 423 | 9054177 | ||

Lca5l
Genomic Map