siRNA Details
| Gene Symbol: Tmtc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198967.5 | NP_945318.2 | 13, 14 | 2113 | ||
| GenBank mRNA | AK046396.1 | - | 12, 13 | 2000 | ||
| AK137545.1 | - | 13, 14 | 2085 | |||
| BC023818.1 | - | 2, 3 | 274 | |||
| BC145513.1 | - | 11, 12 | 1445 | |||

Tmtc1
Genomic Map