siRNA Details
| Gene Symbol: Tmtc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198967.5 | NP_945318.2 | 20 | 2946 | ||
| GenBank mRNA | AK046396.1 | - | 19 | 2833 | ||
| AK137545.1 | - | 20 | 2918 | |||
| BC023818.1 | - | 9 | 1107 | |||
| BC145513.1 | - | 18 | 2278 | |||

Tmtc1
Genomic Map