siRNA Details
| Gene Symbol: Tas2r102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199153.2 | NP_954604.2 | 1 | 862 | ||
| GenBank mRNA | BC117728.2 | - | 1 | 861 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r102| Gene Symbol: Tas2r102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199153.2 | NP_954604.2 | 1 | 862 | ||
| GenBank mRNA | BC117728.2 | - | 1 | 861 | ||