siRNA Details
Gene Symbol: Tas2r143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001452.1 | NP_001001452.1 | 1 | 865 | ||
GenBank mRNA | BC111103.1 | - | 1 | 864 | ||
BC148239.1 | - | 1 | 865 | 40050382 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tas2r143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001452.1 | NP_001001452.1 | 1 | 865 | ||
GenBank mRNA | BC111103.1 | - | 1 | 864 | ||
BC148239.1 | - | 1 | 865 | 40050382 |