siRNA Details
| Gene Symbol: Ugt1a7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201642.4 | NP_964004.1 | 1 | 519 | ||
| GenBank mRNA | AK128918.1 | - | 1 | 445 | ||
| AK144599.1 | - | 1 | 411 | |||
| AK153157.1 | - | 1 | 409 | |||
| AK154973.1 | - | 1 | 442 | |||
| AK159496.1 | - | 1 | 457 | |||
| AK162317.1 | - | 1 | 443 | |||
| AK165465.1 | - | 1 | 443 | |||
| BC141205.1 | - | 1 | 428 | 9055817 | ||
| BC141206.1 | - | 1 | 428 | 9055818 | ||

Ugt1a7c
Genomic Map