siRNA Details
| Gene Symbol: Ugt1a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201643.2 | NP_964005.2 | 1 | 400 | ||
| GenBank mRNA | AB094481.1 | - | 1 | 379 | ||
| AY227196.1 | - | 1 | 400 | |||
| BC139093.1 | - | 1 | 380 | 8862115 | ||
| BC139094.1 | - | 1 | 380 | 8862116 | ||

Ugt1a5
Genomic Map