siRNA Details
| Gene Symbol: Mettl21e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207281.3 | NP_997164.1 | 2 | 179 | ||
| GenBank mRNA | AK029331.1 | - | 2 | 180 | ||
| BC100531.1 | - | 2 | 122 | 30292619 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mettl21e| Gene Symbol: Mettl21e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207281.3 | NP_997164.1 | 2 | 179 | ||
| GenBank mRNA | AK029331.1 | - | 2 | 180 | ||
| BC100531.1 | - | 2 | 122 | 30292619 | ||