siRNA Details
| Gene Symbol: Ano7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271884.1 | NP_001258813.1 | 12 | 1257 | ||
| NM_207031.2 | NP_996914.1 | 12 | 1257 | |||
| GenBank mRNA | BC116706.1 | - | 12 | 1223 | 40096570 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ano7| Gene Symbol: Ano7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271884.1 | NP_001258813.1 | 12 | 1257 | ||
| NM_207031.2 | NP_996914.1 | 12 | 1257 | |||
| GenBank mRNA | BC116706.1 | - | 12 | 1223 | 40096570 | |