siRNA Details
Gene Symbol: Ano7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271884.1 | NP_001258813.1 | 19 | 2087 | ||
NM_207031.2 | NP_996914.1 | 19 | 2087 | |||
GenBank mRNA | BC116706.1 | - | 19 | 2053 | 40096570 |
