siRNA Details
| Gene Symbol: Krt77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003667.1 | NP_001003667.1 | 1 | 19 | ||
| GenBank mRNA | AK132513.1 | - | 1 | 107 | ||
| BC107395.2 | - | 1 | 19 | 40058152 | ||
| BC107396.2 | - | 1 | 19 | 40058154 | ||
| BC141259.1 | - | 1 | 42 | 9055871 | ||

Krt77
Genomic Map